Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DHPS | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001206974.1 | NP_001193903.1 | 2 | 284 | ||
NM_001930.3 | NP_001921.1 | 2 | 433 | |||
NM_013406.2 | NP_037538.1 | 2 | 433 | |||
NR_038192.1 | 2 | 433 | ||||
XM_011527770.1 | XP_011526072.1 | 2 | 433 | |||
XM_011527771.2 | XP_011526073.1 | 2 | 433 | |||
GenBank mRNA | AK291553.1 | - | 2 | 414 | ||
AK304780.1 | - | 2 | 414 | |||
AL520040.3 | - | 2 | 209 | |||
AY336090.1 | - | 2 | 322 | |||
BC000333.2 | - | 2 | 380 | 2822554 | ||
BC014016.2 | - | 2 | 395 | 4129051 | ||
BU844677.1 | - | 2 | 412 | |||
DA524549.1 | - | 2 | 434 | |||
L39068.1 | - | 2 | 349 | |||
U40579.1 | - | 2 | 395 | |||
U79262.1 | - | 2 | 407 |