Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DOCK1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001290223.1 | NP_001277152.1 | 43, 44 | 4531 | ||
NM_001380.4 | NP_001371.1 | 43, 44 | 4468 | |||
XM_011539422.2 | XP_011537724.1 | 43, 44 | 4509 | |||
XM_011539424.1 | XP_011537726.1 | 43, 44 | 4419 | |||
XM_017015813.1 | XP_016871302.1 | 43, 44 | 4441 | |||
XM_017015816.1 | XP_016871305.1 | 26, 27 | 2689 | |||
GenBank mRNA | BC146857.1 | - | 43, 44 | 4421 | 9056785 | |
BX648342.1 | - | 15, 16 | 2556 | |||
D50857.1 | - | 43, 44 | 4382 |