Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other ELAVL4 SIRNA Assays ›
Gene Symbol:
ELAVL4
Gene Name:
ELAV like neuron-specific RNA binding protein 4
Gene Aliases:
HUD, PNEM
Chromosome Location:
Chr.1: 50048014 - 50203786 on Build GRCh38
UniGene ID:
Hs.213050
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
s223476
Purification
HPLC
Standard
HPLC-IVR
Size
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
Availability
Inventoried
Catalog #
4392420
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: ELAVL4
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001144774.2
NP_001138246.1
2, 3
514
NM_001144775.2
NP_001138247.2
2, 3
364
NM_001144776.2
NP_001138248.1
2, 3
326
NM_001144777.2
NP_001138249.1
2, 3
404
NM_001294348.1
NP_001281277.1
2, 3
584
NM_001324208.1
NP_001311137.1
2, 3
404
NM_001324209.1
NP_001311138.1
3, 4
601
NM_001324212.1
NP_001311141.1
3, 4
601
NM_001324213.1
NP_001311142.1
2, 3
584
NM_001324214.1
NP_001311143.1
3, 4
671
NM_001324215.1
NP_001311144.1
2, 3
584
NM_001324216.1
NP_001311145.1
2, 3
514
NM_001324217.1
NP_001311146.1
2, 3
584
NM_021952.4
NP_068771.2
2, 3
514
NR_136725.1
3, 4
671
XM_006710411.3
XP_006710474.1
2, 3
496
XM_011540889.2
XP_011539191.1
2, 3
379
XM_011540890.2
XP_011539192.1
2, 3
378
XM_011540893.2
XP_011539195.1
2, 3
326
XM_011540894.2
XP_011539196.1
3, 4
668
XM_011540895.2
XP_011539197.1
3, 4
572
XM_017000538.1
XP_016856027.1
2, 3
498
XM_017000539.1
XP_016856028.1
2, 3
1125
XM_017000540.1
XP_016856029.1
2, 3
382
XM_017000541.1
XP_016856030.1
2, 3
514
XM_017000542.1
XP_016856031.1
2, 3
363
GenBank mRNA
AK297338.1
-
2, 3
404
AK298817.1
-
2, 3
393
AK298902.1
-
2, 3
584
AK309653.1
-
2, 3
359
AY033995.1
-
2, 3
326
AY033996.1
-
2, 3
337
AY033997.1
-
2, 3
501
AY033998.1
-
2, 3
468
BC036071.1
-
2, 3
472
5286347
BM509527.1
-
2, 3
371
DC351092.1
-
2, 3
404
M62843.1
-
2, 3
338
Back To Top
More Information
Other Species Targeted
This siRNA also targets the following transcripts in Rat: NM_001077651, XM_006238593, XM_006238592, XM_006238591, XM_006238595, and others.
Panther Classification:
Molecular Function -
translation initiation factor
Gene Ontology Categories:
Function(s)
Process(es)
RNA processing
mRNA processing
nucleotide binding
RNA binding
mRNA 3'-UTR binding
AU-rich element binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-green-67fd5c5bdd-gnqd7:80/10.239.161.169:80.
git-commit: 7fb76fe7d0d673187a4c374cd16a9abd242c4866
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.16.3-offline