Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: EPHA1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_005232.4 | NP_005223.4 | 7, 8 | 1560 | ||
XM_006715880.3 | XP_006715943.1 | 6, 7 | 1188 | |||
GenBank mRNA | AK290351.1 | - | 7, 8 | 1524 | ||
AK313263.1 | - | 7, 8 | 1533 | |||
BC130291.1 | - | 7, 8 | 1544 | 40146322 | ||
BC144156.1 | - | 6, 7 | 1696 | |||
CD653381.1 | - | 2, 3 | 449 | |||
M18391.1 | - | 7, 8 | 1566 | |||
Z27409.1 | - | 3, 4 | 618 |