Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PDILT | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_174924.1 | NP_777584.1 | 5 | 796 | ||
XM_011545764.1 | XP_011544066.1 | 5 | 827 | |||
XM_011545765.1 | XP_011544067.1 | 5 | 827 | |||
GenBank mRNA | AK097476.1 | - | 5 | 822 | ||
BC042607.1 | - | 5 | 796 | 5169707 | ||
BC044936.1 | - | 5 | 796 | 5169707 |