Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: EPHA8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001006943.1 | NP_001006944.1 | 3 | 336 | ||
XM_011540969.1 | XP_011539271.1 | 5 | 499 | |||
XM_011540970.1 | XP_011539272.1 | 3 | 233 | |||
XM_011540972.1 | XP_011539274.1 | 4 | 400 | |||
XM_011540973.1 | XP_011539275.1 | 2 | 135 | |||
GenBank mRNA | BC038796.1 | - | 3 | 266 | ||
BC072417.1 | - | 3 | 336 | |||
DR007053.1 | - | 3 | 346 |