Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ABCD1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000033.3 | NP_000024.2 | 1 | 1035 | ||
GenBank mRNA | AY421736.1 | - | 1 | 166 | ||
BC015541.1 | - | 1 | 1016 | 3896490 | ||
BC025358.1 | - | 1 | 1035 | 4907640 | ||
Z21876.1 | - | 1 | 1022 |