Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other PUF60 SIRNA Assays ›
Gene Symbol:
PUF60
Gene Name:
poly(U) binding splicing factor 60
Gene Aliases:
FIR, RoBPI, SIAHBP1, VRJS
Chromosome Location:
Chr.8: 143816344 - 143829386 on Build GRCh38
UniGene ID:
Hs.521924
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
s223539
Purification
HPLC
Standard
HPLC-IVR
Size
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
Availability
Inventoried
Catalog #
4392420
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: PUF60
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001136033.2
NP_001129505.1
6
580
NM_001271096.1
NP_001258025.1
5
507
NM_001271097.1
NP_001258026.1
4
423
NM_001271098.1
NP_001258027.1
6
558
NM_001271099.1
NP_001258028.1
5
474
NM_001271100.1
NP_001258029.1
5
529
NM_014281.4
NP_055096.2
5
510
NM_078480.2
NP_510965.1
6
561
XM_011516929.2
XP_011515231.1
7
760
XM_011516930.2
XP_011515232.1
6
611
XM_017013234.1
XP_016868723.1
6
2093
XM_017013235.1
XP_016868724.1
6
663
XM_017013236.1
XP_016868725.1
5
2042
XM_017013237.1
XP_016868726.1
7
822
XM_017013238.1
XP_016868727.1
7
622
XM_017013239.1
XP_016868728.1
6
588
XM_017013240.1
XP_016868729.1
5
537
GenBank mRNA
AF114818.1
-
6
542
AF190744.1
-
6
470
AF217197.2
-
5
437
AK055941.1
-
5
1164
AK292373.1
-
6
542
AL522753.3
-
6
508
BC008875.2
-
5
476
4138778
BC009734.1
-
5
459
3877563
BC011265.1
-
6
497
4155549
BC011979.1
-
4
376
3863660
BC035435.2
-
5
461
BG771821.1
-
5
528
BX418925.2
-
5
459
CN371576.1
-
5
477
CR457143.1
-
6
478
CT000277.1
-
5
442
CX163878.1
-
4
381
U51586.1
-
4
424
Back To Top
More Information
Panther Classification:
Molecular Function -
RNA splicing factor
Gene Ontology Categories:
Function(s)
Process(es)
transcription, DNA-templated
regulation of transcription, DNA-templated
mRNA processing
apoptotic process
RNA splicing
cell-cell adhesion
nucleotide binding
DNA binding
protein binding
identical protein binding
poly(A) RNA binding
cadherin binding involved in cell-cell adhesion
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-7487cff77c-nnwcq:80/10.239.162.212:80.
git-commit: c531a7a49d92b854cb004d8acd820c41186dcba6
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.16.2-2024.09.38-1.0