Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FOXE1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_004473.3 | NP_004464.2 | 1 | 2200 | ||
GenBank mRNA | U89995.1 | - | 1 | 2237 |