Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TTLL5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_015072.4 | NP_055887.3 | 20 | 2192 | ||
GenBank mRNA | AB023215.1 | - | 18 | 1752 | ||
AK309748.1 | - | 21 | 2174 | |||
AL136808.1 | - | 6 | 640 | |||
AY237126.1 | - | 20 | 2122 | |||
BC016625.1 | - | 7 | 773 | |||
BC136472.1 | - | 20 | 2082 | 9020461 | ||
BC136473.1 | - | 20 | 2082 | 9020462 | ||
BX647530.1 | - | 13 | 1223 | |||
CR533554.1 | - | 5 | 570 |