Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: AKR1B1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001628.2 | NP_001619.1 | 6 | 659 | ||
XM_005250234.3 | XP_005250291.1 | 6 | 668 | |||
GenBank mRNA | AB209796.1 | - | 6 | 578 | ||
AF328729.1 | - | 6 | 600 | |||
AK313439.1 | - | 6 | 650 | |||
BC000260.1 | - | 6 | 659 | 3357652 | ||
BC005387.1 | - | 6 | 623 | 3960811 | ||
BC010391.1 | - | 6 | 625 | 4049736 | ||
BT019859.1 | - | 6 | 585 | |||
CR450351.1 | - | 6 | 585 | |||
CR542203.1 | - | 6 | 585 | |||
J04795.1 | - | 6 | 594 | |||
J05017.1 | - | 6 | 621 | |||
J05474.1 | - | 6 | 617 | |||
M34720.1 | - | 6 | 598 | |||
X15414.1 | - | 6 | 630 |