Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FNTA | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_002027.2 | NP_002018.1 | 8, 9 | 1065 | ||
NR_033698.1 | 7, 8 | 979 | ||||
GenBank mRNA | AB209689.1 | - | 2, 3 | 6729 | ||
AK091717.1 | - | 3, 4 | 1908 | |||
AK123002.1 | - | 8, 9 | 1452 | |||
AK292121.1 | - | 8, 9 | 1041 | |||
BC017029.2 | - | 8, 9 | 1008 | |||
BC084566.1 | - | 8, 9 | 1025 | 6423445 | ||
BT009854.1 | - | 8, 9 | 1017 | |||
BX648381.1 | - | 3, 4 | 2836 | |||
L00634.1 | - | 8, 9 | 1033 | |||
L10413.1 | - | 8, 9 | 1023 |