Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FNTA | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_002027.2 | NP_002018.1 | 8 | 911 | ||
NR_033698.1 | 7 | 825 | ||||
GenBank mRNA | AB209689.1 | - | 2 | 6575 | ||
AK091717.1 | - | 3 | 1754 | |||
AK123002.1 | - | 8 | 1298 | |||
AK292121.1 | - | 8 | 887 | |||
BC017029.2 | - | 8 | 854 | |||
BC084566.1 | - | 8 | 871 | 6423445 | ||
BT009854.1 | - | 8 | 863 | |||
BX648381.1 | - | 3 | 2682 | |||
L00634.1 | - | 8 | 879 | |||
L10413.1 | - | 8 | 869 |