Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DDAH1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001134445.1 | NP_001127917.1 | 4 | 630 | ||
NM_012137.3 | NP_036269.1 | 3 | 598 | |||
XM_005270707.2 | XP_005270764.1 | 3 | 307 | |||
XM_005270710.2 | XP_005270767.1 | 3 | 526 | |||
XM_011541158.1 | XP_011539460.1 | 5 | 630 | |||
XM_017000889.1 | XP_016856378.1 | 3 | 284 | |||
XM_017000890.1 | XP_016856379.1 | 3 | 2036 | |||
GenBank mRNA | AK098405.1 | - | 4 | 452 | ||
AK294752.1 | - | 3 | 223 | |||
AK302531.1 | - | 3 | 251 | |||
AK304876.1 | - | 4 | 634 | |||
AK309225.1 | - | 3 | 207 | |||
AK312432.1 | - | 3 | 559 | |||
BC033680.1 | - | 3 | 536 | 5189970 | ||
BC043235.2 | - | 3 | 320 | |||
BX648145.1 | - | 4 | 517 | |||
EU794605.1 | - | 3 | 536 |