Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SLC7A11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_014331.3 | NP_055146.1 | 11, 12 | 1734 | ||
GenBank mRNA | AB026891.1 | - | 11, 12 | 1689 | ||
AF200708.1 | - | 11, 12 | 1699 | |||
AF252872.1 | - | 11, 12 | 1685 | |||
AJ277882.1 | - | 11, 12 | 1454 | |||
AK290359.1 | - | 11, 12 | 1734 | |||
AK314855.1 | - | 11, 12 | 1628 | |||
BC012087.1 | - | 11, 12 | 1614 | 4562994 |