Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MSRB3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001031679.2 | NP_001026849.1 | 4 | 498 | ||
NM_001193460.1 | NP_001180389.1 | 5 | 662 | |||
NM_001193461.1 | NP_001180390.1 | 4 | 360 | |||
NM_198080.3 | NP_932346.1 | 3 | 371 | |||
GenBank mRNA | AK293084.1 | - | 4 | 468 | ||
AK299065.1 | - | 3 | 310 | |||
AK316365.1 | - | 3 | 342 | |||
AL833622.1 | - | 4 | 416 | |||
AY358229.1 | - | 3 | 306 | |||
BC040053.1 | - | 5 | 563 | 6025598 | ||
BX500552.1 | - | 4 | 362 | |||
BX648776.1 | - | 4 | 417 | |||
DC357690.1 | - | 3 | 371 |