Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ARIH1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_005744.3 | NP_005735.2 | 13 | 1879 | ||
XM_011521438.2 | XP_011519740.1 | 12 | 1855 | |||
XM_011521439.2 | XP_011519741.1 | 14 | 1393 | |||
GenBank mRNA | AB014774.1 | - | 4 | 439 | ||
AB209432.1 | - | 14 | 1508 | |||
AF072832.1 | - | 13 | 1565 | |||
AJ009771.1 | - | 6 | 675 | |||
AJ130976.1 | - | 13 | 1283 | |||
AJ243190.1 | - | 13 | 1879 | |||
AK312715.1 | - | 13 | 1878 | |||
BC051877.1 | - | 13 | 1814 | 5742498 | ||
BX641117.1 | - | 1 | 429 |