Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: WHRN | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001083885.2 | NP_001077354.2 | 12 | 2169 | ||
NM_001173425.1 | NP_001166896.1 | 12 | 3330 | |||
NM_015404.3 | NP_056219.3 | 12 | 3333 | |||
XM_011518484.1 | XP_011516786.1 | 13 | 3419 | |||
XM_011518485.1 | XP_011516787.1 | 14 | 3267 | |||
XM_011518486.1 | XP_011516788.1 | 13 | 3416 | |||
XM_011518487.1 | XP_011516789.1 | 12 | 3293 | |||
XM_011518488.1 | XP_011516790.1 | 11 | 3176 | |||
XM_011518495.1 | XP_011516797.1 | 10 | 1671 | |||
GenBank mRNA | AB040959.1 | - | 12 | 2847 | ||
AK022854.1 | - | 8 | 2047 | |||
AL110228.1 | - | 12 | 2127 | |||
BC011918.2 | - | 3 | 442 | |||
BC014524.1 | - | 3830 | ||||
BC136416.1 | - | 12 | 3079 | 9020404 | ||
BC142614.1 | - | 12 | 3333 | 40147287 | ||
BC142684.1 | - | 12 | 3308 | 40148465 |