Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: OIP5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001317860.1 | NP_001304789.1 | 2 | 433 | ||
NM_007280.1 | NP_009211.1 | 2 | 433 | |||
GenBank mRNA | AF025441.1 | - | 2 | 379 | ||
AW406383.1 | - | 2 | 300 | |||
BC015050.1 | - | 2 | 433 | 3908972 |