Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MTG2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_015666.3 | NP_056481.1 | 3 | 377 | ||
XM_005260393.1 | XP_005260450.1 | 3 | 367 | |||
XM_006723774.3 | XP_006723837.1 | 4 | 461 | |||
XM_011528776.1 | XP_011527078.1 | 3 | 2381 | |||
XM_017027805.1 | XP_016883294.1 | 2 | 2531 | |||
XM_017027806.1 | XP_016883295.1 | 3 | 366 | |||
XM_017027807.1 | XP_016883296.1 | 2 | 2531 | |||
XM_017027808.1 | XP_016883297.1 | 3 | 367 | |||
GenBank mRNA | AK001603.1 | - | 2 | 357 | ||
AK124572.1 | - | 3 | 2668 | |||
AK299147.1 | - | 2 | 151 | |||
AK299189.1 | - | 3 | 360 | |||
AK299726.1 | - | 3 | 360 | |||
AK316027.1 | - | 2 | 157 | |||
AK316325.1 | - | 2 | 154 | |||
BC012073.1 | - | 2 | 358 | |||
BC036716.2 | - | 2 | 351 | 4900170 | ||
DA970672.1 | - | 3 | 377 |