Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PHGDH | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_006623.3 | NP_006614.2 | 4, 5 | 642 | ||
XM_011541226.1 | XP_011539528.1 | 6, 7 | 923 | |||
XM_011541227.2 | XP_011539529.1 | 6, 7 | 628 | |||
XM_011541228.1 | XP_011539530.1 | 6, 7 | 541 | |||
XM_011541231.2 | XP_011539533.1 | 6, 7 | 745 | |||
GenBank mRNA | AF171236.1 | - | 1, 2 | 32 | ||
AF171237.1 | - | 4, 5 | 488 | |||
AK093306.1 | - | 3, 4 | 1820 | |||
AK309662.1 | - | 6, 7 | 836 | |||
AK310132.1 | - | 5, 6 | 593 | |||
AK315360.1 | - | 4, 5 | 563 | |||
BC000303.2 | - | 4, 5 | 482 | 2820369 | ||
BC001349.2 | - | 4, 5 | 477 | 2961389 | ||
BC011262.1 | - | 4, 5 | 551 | 4156703 | ||
BC023235.1 | - | 4, 5 | 455 | |||
CR456795.1 | - | 4, 5 | 415 | |||
FJ224347.1 | - | 1107 |