Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PHGDH | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_006623.3 | NP_006614.2 | 2 | 507 | ||
XM_011541226.1 | XP_011539528.1 | 2 | 566 | |||
XM_011541227.2 | XP_011539529.1 | 2 | 271 | |||
XM_011541228.1 | XP_011539530.1 | 2 | 184 | |||
XM_011541231.2 | XP_011539533.1 | 2 | 368 | |||
GenBank mRNA | AF171237.1 | - | 2 | 353 | ||
AK093306.1 | - | 1 | 1685 | |||
AK309662.1 | - | 2 | 367 | |||
AK310132.1 | - | 2 | 367 | |||
AK315360.1 | - | 2 | 428 | |||
BC000303.2 | - | 2 | 347 | 2820369 | ||
BC001349.2 | - | 2 | 342 | 2961389 | ||
BC011262.1 | - | 2 | 416 | 4156703 | ||
BC023235.1 | - | 2 | 320 | |||
CR456795.1 | - | 2 | 280 | |||
DA731339.1 | - | 2 | 588 | |||
FJ224347.1 | - | 972 |