Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PHGDH | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_006623.3 | NP_006614.2 | 5, 6 | 738 | ||
XM_011541226.1 | XP_011539528.1 | 7, 8 | 1019 | |||
XM_011541227.2 | XP_011539529.1 | 7, 8 | 724 | |||
XM_011541228.1 | XP_011539530.1 | 7, 8 | 637 | |||
XM_011541231.2 | XP_011539533.1 | 7, 8 | 841 | |||
GenBank mRNA | AF171236.1 | - | 2, 3 | 128 | ||
AF171237.1 | - | 5, 6 | 584 | |||
AK093306.1 | - | 4, 5 | 1916 | |||
AK309662.1 | - | 7, 8 | 932 | |||
AK310132.1 | - | 6, 7 | 689 | |||
AK315360.1 | - | 5, 6 | 659 | |||
BC000303.2 | - | 5, 6 | 578 | 2820369 | ||
BC001349.2 | - | 5, 6 | 573 | 2961389 | ||
BC011262.1 | - | 5, 6 | 647 | 4156703 | ||
BC023235.1 | - | 5, 6 | 551 | |||
CR456795.1 | - | 5, 6 | 511 | |||
FJ224347.1 | - | 1203 |