Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HIBCH | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_014362.3 | NP_055177.2 | 2 | 351 | ||
NM_198047.2 | NP_932164.1 | 2 | 351 | |||
XM_011510953.1 | XP_011509255.1 | 2 | 300 | |||
GenBank mRNA | AK222979.1 | - | 2 | 75 | ||
AK223023.1 | - | 2 | 75 | |||
AK308042.1 | - | 2 | 75 | |||
AK313651.1 | - | 2 | 81 | |||
BC005190.1 | - | 2 | 128 | |||
BC067822.1 | - | 2 | 80 | 5266813 | ||
BI764401.1 | - | 2 | 161 | |||
BP193393.1 | - | 2 | 351 | |||
CA453768.1 | - | 132 | ||||
DB034438.1 | - | 2 | 301 | |||
U66669.1 | - | 2 | 104 |