Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: BLOC1S1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001487.3 | NP_001478.2 | 2 | 182 | ||
NR_037656.1 | 2 | 445 | ||||
GenBank mRNA | AK307069.1 | - | 2 | 178 | ||
BC130640.1 | - | 2 | 147 | 40146671 | ||
BC132795.1 | - | 2 | 147 | 40146817 | ||
BQ899204.1 | - | 2 | 445 | |||
BU601406.1 | - | 2 | 192 | |||
D64007.1 | - | 2 | 178 | |||
DA243440.1 | - | 2 | 180 | |||
S82447.1 | - | 2 | 135 |
Gene Symbol: BLOC1S1-RDH5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_037658.1 | 2 | 182 | |||
GenBank mRNA | AK096656.1 | - | 2 | 164 | ||
BG722558.1 | - | 2 | 149 |