Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PPA2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_006903.4 | NP_008834.3 | 7 | 686 | ||
NM_176866.2 | NP_789842.2 | 4 | 467 | |||
NM_176867.3 | NP_789843.2 | 2 | 275 | |||
NM_176869.2 | NP_789845.1 | 8 | 773 | |||
GenBank mRNA | AB026722.1 | - | 7 | 680 | ||
AF161473.1 | - | 9 | 762 | |||
AF217187.1 | - | 8 | 767 | |||
AK000466.1 | - | 8 | 765 | |||
AK289607.1 | - | 7 | 664 | |||
AK299125.1 | - | 8 | 762 | |||
AL136655.1 | - | 9 | 745 | |||
BC022803.1 | - | 2 | 251 | |||
BC035569.1 | - | 8 | 745 | |||
BC039462.1 | - | 4 | 414 | |||
BC057219.1 | - | 7 | 679 | 4289401 |