Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HPGDS | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_014485.2 | NP_055300.1 | 6 | 572 | ||
XM_005262932.2 | XP_005262989.1 | 5 | 479 | |||
GenBank mRNA | AK290075.1 | - | 6 | 572 | ||
BC020734.1 | - | 6 | 549 | 4731411 | ||
CR541662.1 | - | 5 | 481 | |||
CR541679.1 | - | 5 | 481 | |||
D82073.1 | - | 5 | 491 |