Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GLDC | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000170.2 | NP_000161.2 | 9 | 1416 | ||
GenBank mRNA | AK310534.1 | - | 5 | 791 | ||
AK314156.1 | - | 9 | 1418 | |||
BC111993.1 | - | 9 | 1289 | 8327461 | ||
BC111995.1 | - | 9 | 1289 | 8327463 | ||
D90239.1 | - | 9 | 1373 | |||
M63635.1 | - | 9 | 1304 | |||
M64590.1 | - | 9 | 1373 |