Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HTRA2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001321727.1 | NP_001308656.1 | 1 | 898 | ||
NM_001321728.1 | NP_001308657.1 | 1 | 898 | |||
NM_013247.4 | NP_037379.1 | 1 | 898 | |||
NM_145074.2 | NP_659540.1 | 1 | 898 | |||
NR_135769.1 | 1 | 898 | ||||
GenBank mRNA | AF020760.2 | - | 1 | 313 | ||
AF141305.1 | - | 1 | 898 | |||
AF141306.1 | - | 1 | 257 | |||
AF141307.1 | - | 1 | 134 | |||
AF184911.1 | - | 1 | 313 | |||
AK225932.1 | - | 1 | 331 | |||
AK291977.1 | - | 1 | 317 | |||
BC000096.2 | - | 1 | 324 | 3508944 |