Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SLC9C1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001320531.1 | NP_001307460.1 | 14 | 1858 | ||
NM_183061.2 | NP_898884.1 | 15 | 2002 | |||
NR_135297.1 | 11 | 1443 | ||||
XM_006713603.2 | XP_006713666.1 | 15 | 2037 | |||
XM_011512718.1 | XP_011511020.1 | 14 | 1840 | |||
XM_011512719.1 | XP_011511021.1 | 15 | 2002 | |||
XM_011512720.1 | XP_011511022.1 | 15 | 2002 | |||
XM_011512721.1 | XP_011511023.1 | 16 | 2180 | |||
XM_011512724.1 | XP_011511026.1 | 15 | 2002 | |||
XM_011512725.2 | XP_011511027.1 | 5 | 551 | |||
XM_011512726.2 | XP_011511028.1 | 15 | 2002 | |||
XM_011512727.2 | XP_011511029.1 | 15 | 2002 | |||
XM_017006246.1 | XP_016861735.1 | 13 | 1705 | |||
XM_017006247.1 | XP_016861736.1 | 15 | 2002 | |||
XM_017006248.1 | XP_016861737.1 | 15 | 2002 | |||
XM_017006249.1 | XP_016861738.1 | 15 | 2002 | |||
XM_017006250.1 | XP_016861739.1 | 15 | 2002 | |||
GenBank mRNA | AK128084.1 | - | 15 | 2002 | ||
BC044801.1 | - | 14 | 1827 | 4827529 | ||
BC143774.1 | - | 10 | 1288 |