Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GRIK4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001282470.2 | NP_001269399.1 | 19 | 2824 | ||
NM_014619.4 | NP_055434.2 | 20 | 2784 | |||
XM_011542784.1 | XP_011541086.1 | 18 | 2480 | |||
XM_011542786.2 | XP_011541088.1 | 17 | 3284 | |||
XM_011542787.2 | XP_011541089.1 | 13 | 3734 | |||
XM_017017621.1 | XP_016873110.1 | 18 | 2468 | |||
XM_017017622.1 | XP_016873111.1 | 18 | 2438 | |||
GenBank mRNA | AK292726.1 | - | 20 | 2771 | ||
AK314284.1 | - | 20 | 2826 | |||
S67803.1 | - | 18 | 2484 |