Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: BRF1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001242786.1 | NP_001229715.1 | 7 | 789 | ||
NM_001242787.1 | NP_001229716.1 | 7 | 789 | |||
NM_001242788.1 | NP_001229717.1 | 6 | 999 | |||
NM_001242789.1 | NP_001229718.1 | 2 | 279 | |||
NM_001519.3 | NP_001510.2 | 7 | 1080 | |||
NM_145685.2 | NP_663718.1 | 3 | 988 | |||
XM_005267561.3 | XP_005267618.1 | 7 | 1102 | |||
XM_005267563.3 | XP_005267620.1 | 3 | 1792 | |||
XM_006720123.2 | XP_006720186.1 | 2 | 1220 | |||
XM_011536672.2 | XP_011534974.1 | 7 | 1102 | |||
XM_011536673.1 | XP_011534975.1 | 6 | 952 | |||
XM_011536674.1 | XP_011534976.1 | 3 | 1370 | |||
GenBank mRNA | AJ297406.1 | - | 3 | 1542 | ||
AJ297407.1 | - | 3 | 996 | |||
AK096471.1 | - | 2 | 279 | |||
AK294899.1 | - | 7 | 789 | |||
AK295579.1 | - | 7 | 702 | |||
BC071637.1 | - | 2 | 144 | |||
BC086856.1 | - | 6 | 732 | 6166751 | ||
BC103859.1 | - | 3 | 501 | |||
BC103860.2 | - | 2 | 146 | |||
BC103861.2 | - | 3 | 296 | |||
BC133655.1 | - | 3 | 296 | |||
U28838.1 | - | 7 | 1083 | |||
U75276.1 | - | 7 | 784 |