Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TWF2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_007284.3 | NP_009215.1 | 6, 7 | 764 | ||
GenBank mRNA | AF109365.1 | - | 6, 7 | 665 | ||
AL136773.1 | - | 6, 7 | 731 | |||
BC000327.2 | - | 6, 7 | 715 | 2822004 | ||
BC003161.1 | - | 6, 7 | 681 | 2957960 | ||
BC016452.1 | - | 6, 7 | 731 | 3452300 | ||
CR456732.1 | - | 6, 7 | 613 | |||
CR533520.1 | - | 6, 7 | 613 | |||
Y17169.1 | - | 6, 7 | 717 |