Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PILRA | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_013439.2 | NP_038467.2 | 6, 7 | 996 | ||
NM_178272.1 | NP_840056.1 | 5, 6 | 777 | |||
NM_178273.1 | NP_840057.1 | 4, 5 | 743 | |||
GenBank mRNA | AF161080.1 | - | 6, 7 | 996 | ||
AJ400841.1 | - | 6, 7 | 937 | |||
AJ400842.1 | - | 5, 6 | 694 | |||
AJ400843.1 | - | 4, 5 | 660 |