Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PACSIN1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001199583.2 | NP_001186512.1 | 8, 9 | 1299 | ||
NM_020804.4 | NP_065855.1 | 8, 9 | 1312 | |||
XM_011514541.1 | XP_011512843.1 | 8, 9 | 1167 | |||
GenBank mRNA | AB037800.2 | - | 8, 9 | 1269 | ||
AF242529.1 | - | 8, 9 | 1235 | |||
AK289956.1 | - | 8, 9 | 1297 | |||
AK294132.1 | - | 8, 9 | 1299 | |||
AL834211.1 | - | 8, 9 | 1245 | |||
BC040228.1 | - | 8, 9 | 1284 | 5173129 | ||
BC128219.1 | - | 5, 6 | 692 |