Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HNRNPA1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_002136.3 | NP_002127.1 | 10 | 1613 | ||
NM_031157.3 | NP_112420.1 | 11 | 1769 | |||
XM_017019251.1 | XP_016874740.1 | 9 | 1532 | |||
GenBank mRNA | AK129814.1 | - | 10 | 1555 | ||
BC002355.2 | - | 10 | 1534 | 2821751 | ||
BC009600.1 | - | 10 | 1556 | 4134638 | ||
BC012158.1 | - | 10 | 1516 | 4564655 | ||
BC033714.1 | - | 10 | 1545 | 4420039 | ||
BC035253.1 | - | 6 | 972 | |||
BC052296.1 | - | 10 | 1525 | 6297642 | ||
BC074502.1 | - | 10 | 1538 | 3455742 | ||
BC103707.1 | - | 9 | 1364 | 6154800 | ||
X06747.1 | - | 10 | 1579 |
Gene Symbol: XKRX | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_011530954.2 | XP_011529256.1 | 4 | 3972 |