Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: AIRE | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000383.3 | NP_000374.1 | 14 | 1928 | ||
GenBank mRNA | AB006682.1 | - | 14 | 1886 | ||
AB006683.1 | - | 8 | 1404 | |||
AB006685.1 | - | 9 | 1322 | |||
BC137268.1 | - | 7 | 1585 | 9021265 | ||
BC137270.1 | - | 7 | 1584 | 9021267 | ||
Z97990.1 | - | 14 | 1879 |