Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TMEM50B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_006134.6 | NP_006125.2 | 5 | 566 | ||
NR_040016.1 | 5 | 566 | ||||
XM_011529746.1 | XP_011528048.1 | 5 | 564 | |||
GenBank mRNA | AF045606.2 | - | 5 | 511 | ||
AF086280.1 | - | 4 | 400 | |||
AK309977.1 | - | 5 | 493 | |||
AK310701.1 | - | 5 | 422 | |||
AK311870.1 | - | 4 | 386 | |||
AY358634.1 | - | 5 | 490 | |||
AY820138.1 | - | 4 | 353 | |||
BC000569.2 | - | 5 | 482 | 3342648 | ||
DQ286229.1 | - | 4 | 353 |