Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: IFNA1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_024013.2 | NP_076918.1 | 1 | 732 | ||
GenBank mRNA | AB578886.1 | - | 1 | 745 | ||
V00537.1 | - | 1 | 734 | |||
V00538.1 | - | 1 | 732 |