Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RBPJ | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_005349.3 | NP_005340.2 | 12 | 1668 | ||
NM_015874.4 | NP_056958.3 | 11 | 1615 | |||
NM_203283.2 | NP_976028.1 | 11 | 1552 | |||
NM_203284.2 | NP_976029.1 | 12 | 1798 | |||
XM_005248161.3 | XP_005248218.1 | 12 | 1682 | |||
XM_011513840.2 | XP_011512142.1 | 12 | 2074 | |||
XM_017008170.1 | XP_016863659.1 | 11 | 1471 | |||
XM_017008171.1 | XP_016863660.1 | 11 | 1450 | |||
XM_017008172.1 | XP_016863661.1 | 12 | 1613 | |||
XM_017008173.1 | XP_016863662.1 | 12 | 1891 | |||
XM_017008174.1 | XP_016863663.1 | 12 | 1707 | |||
XM_017008175.1 | XP_016863664.1 | 12 | 1623 | |||
GenBank mRNA | AK126357.1 | - | 6 | 4083 | ||
AK302230.1 | - | 11 | 1405 | |||
BC020780.1 | - | 11 | 1552 | 4718839 | ||
BC064976.1 | - | 12 | 1573 | 6152924 | ||
BM560796.1 | - | 1 | 120 | |||
L07872.1 | - | 11 | 1510 |