Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RBPJ | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_005349.3 | NP_005340.2 | 8, 9 | 1016 | ||
NM_015874.4 | NP_056958.3 | 7, 8 | 963 | |||
NM_203283.2 | NP_976028.1 | 7, 8 | 900 | |||
NM_203284.2 | NP_976029.1 | 8, 9 | 1146 | |||
XM_005248161.3 | XP_005248218.1 | 8, 9 | 1030 | |||
XM_011513840.2 | XP_011512142.1 | 8, 9 | 1422 | |||
XM_017008170.1 | XP_016863659.1 | 7, 8 | 819 | |||
XM_017008171.1 | XP_016863660.1 | 7, 8 | 798 | |||
XM_017008172.1 | XP_016863661.1 | 8, 9 | 961 | |||
XM_017008173.1 | XP_016863662.1 | 8, 9 | 1239 | |||
XM_017008174.1 | XP_016863663.1 | 8, 9 | 1055 | |||
XM_017008175.1 | XP_016863664.1 | 8, 9 | 971 | |||
GenBank mRNA | AK126357.1 | - | 2, 3 | 3509 | ||
AK302230.1 | - | 7, 8 | 753 | |||
AK303244.1 | - | 7, 8 | 812 | |||
AK311419.1 | - | 7, 8 | 799 | |||
BC020780.1 | - | 7, 8 | 900 | 4718839 | ||
BC064976.1 | - | 8, 9 | 921 | 6152924 | ||
D14041.1 | - | 5, 6 | 778 | |||
L07872.1 | - | 7, 8 | 858 |