Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: APRT | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000485.2 | NP_000476.1 | 1, 2 | 108 | ||
NM_001030018.1 | NP_001025189.1 | 1, 2 | 108 | |||
GenBank mRNA | BC107151.2 | - | 1, 2 | 98 | 40032160 | |
BM423481.1 | - | 132 | ||||
BM550173.1 | - | 1, 2 | 131 | |||
CR749423.1 | - | 1, 2 | 99 |