Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: IL1B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000576.2 | NP_000567.1 | 4, 5 | 396 | ||
XM_017003988.1 | XP_016859477.1 | 3, 4 | 397 | |||
GenBank mRNA | BC008678.1 | - | 4, 5 | 396 | 3875593 | |
BT007213.1 | - | 3, 4 | 309 | |||
CR407679.1 | - | 3, 4 | 309 | |||
K02770.1 | - | 4, 5 | 395 | |||
M15330.1 | - | 4, 5 | 395 | |||
M54933.1 | - | 4, 5 | 396 | |||
X56087.1 | - | 4, 5 | 366 |