Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: IL1B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000576.2 | NP_000567.1 | 6, 7 | 689 | ||
XM_017003988.1 | XP_016859477.1 | 5, 6 | 690 | |||
GenBank mRNA | AB451494.1 | - | 2, 3 | 254 | ||
AF043335.1 | - | 2, 3 | 254 | |||
AK313983.1 | - | 5, 6 | 1409 | |||
BC008678.1 | - | 6, 7 | 689 | 3875593 | ||
BT007213.1 | - | 5, 6 | 602 | |||
CR407679.1 | - | 5, 6 | 602 | |||
K02770.1 | - | 6, 7 | 688 | |||
M15330.1 | - | 6, 7 | 688 | |||
M54933.1 | - | 6, 7 | 689 | |||
X02532.1 | - | 6, 7 | 679 | |||
X56087.1 | - | 6, 7 | 659 |