Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other IL15RA SIRNA Assays ›
Gene Symbol:
IL15RA
Gene Name:
interleukin 15 receptor subunit alpha
Gene Aliases:
CD215
Chromosome Location:
Chr.10: 5948897 - 5978741 on Build GRCh38
UniGene ID:
Hs.445124
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
s223937
Purification
HPLC
Standard
HPLC-IVR
Size
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
Availability
Inventoried
Catalog #
4392420
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: IL15RA
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001243539.1
NP_001230468.1
6
1004
NM_001256765.1
NP_001243694.1
7
953
NM_002189.3
NP_002180.1
6
800
NM_172200.2
NP_751950.2
5
701
NR_046362.1
4
710
XM_011519461.2
XP_011517763.1
8
1194
XM_011519462.2
XP_011517764.1
7
1112
XM_011519463.2
XP_011517765.1
7
1095
XM_011519464.2
XP_011517766.1
7
1041
XM_011519465.1
XP_011517767.1
8
1194
XM_011519466.2
XP_011517768.1
8
1194
XM_011519467.2
XP_011517769.1
6
957
XM_011519468.1
XP_011517770.1
8
1194
XM_011519469.1
XP_011517771.1
7
1095
XM_011519470.2
XP_011517772.1
5
860
XM_011519471.2
XP_011517773.1
6
925
XM_011519472.2
XP_011517774.1
5
764
XM_011519474.2
XP_011517776.1
5
747
XM_011519475.2
XP_011517777.1
4
665
XM_011519476.2
XP_011517778.1
4
655
XM_011519477.2
XP_011517779.1
4
660
XM_017016195.1
XP_016871684.1
7
1041
XM_017016196.1
XP_016871685.1
6
766
XM_017016197.1
XP_016871686.1
7
781
XM_017016198.1
XP_016871687.1
6
925
XM_017016200.1
XP_016871689.1
5
1352
XM_017016201.1
XP_016871690.1
6
1049
XM_017016202.1
XP_016871691.1
7
770
XM_017016203.1
XP_016871692.1
5
1352
GenBank mRNA
AF035279.1
-
6
735
AK304211.1
-
5
612
BC065506.1
-
4
702
BC074726.2
-
6
712
30915179
BC107777.1
-
6
868
6497994
BC121140.2
-
6
707
40120998
BC121141.2
-
6
712
40120999
BM546106.1
-
4
722
CR457064.1
-
5
573
CR542023.1
-
6
681
HQ401283.1
-
7
834
JX987305.1
-
7
834
JX987306.1
-
7
834
JX987307.1
-
7
834
JX987308.1
-
7
834
JX987309.1
-
7
834
JX987310.1
-
7
834
JX987311.1
-
7
834
U31628.1
-
6
763
Back To Top
More Information
Panther Classification:
Molecular Function -
transmembrane signal receptor
Gene Ontology Categories:
Function(s)
Process(es)
signal transduction
cell proliferation
cytokine-mediated signaling pathway
signal transducer activity
cytokine receptor activity
protein binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-7487cff77c-nnwcq:80/10.239.162.212:80.
git-commit: c531a7a49d92b854cb004d8acd820c41186dcba6
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.16.2-2024.09.38-1.0