Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: AQP9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001320635.1 | NP_001307564.1 | 1 | 432 | ||
NM_020980.4 | NP_066190.2 | 1 | 432 | |||
GenBank mRNA | AB008775.1 | - | 1 | 278 | ||
AF016495.1 | - | 1 | 290 | |||
AK313214.1 | - | 1 | 357 | |||
BC026258.1 | - | 1 | 247 | 4734559 | ||
CR542027.1 | - | 1 | 62 | |||
DC410284.1 | - | 1 | 432 |