Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KPNA1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_002264.3 | NP_002255.3 | 6 | 638 | ||
NR_026698.1 | 7 | 761 | ||||
XM_005247437.3 | XP_005247494.1 | 6 | 520 | |||
XM_005247439.3 | XP_005247496.1 | 4 | 1268 | |||
XM_011512795.2 | XP_011511097.1 | 7 | 572 | |||
XM_011512797.2 | XP_011511099.1 | 7 | 2235 | |||
XM_017006363.1 | XP_016861852.1 | 7 | 2209 | |||
XM_017006364.1 | XP_016861853.1 | 7 | 638 | |||
XM_017006365.1 | XP_016861854.1 | 2 | 902 | |||
GenBank mRNA | AK125626.1 | - | 3 | 588 | ||
AK128280.1 | - | 4 | 1268 | |||
AK312713.1 | - | 7 | 756 | |||
BC002374.2 | - | 6 | 627 | 2822859 | ||
BC003009.1 | - | 6 | 627 | 2822859 | ||
BC090864.1 | - | 6 | 609 | 6173605 | ||
BT006959.1 | - | 5 | 450 | |||
CR456743.1 | - | 5 | 450 | |||
DN991078.1 | - | 7 | 603 | |||
S75295.1 | - | 6 | 496 | |||
U20620.1 | - | 4 | 303 |