Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SP5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001003845.2 | NP_001003845.1 | 2 | 1873 | ||
XM_005246542.4 | XP_005246599.1 | 2 | 3000 | |||
XM_011511159.2 | XP_011509461.1 | 2 | 3036 | |||
GenBank mRNA | AB096175.1 | - | 2 | 1868 |