Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: IYD | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001164694.1 | NP_001158166.1 | 2 | 385 | ||
NM_001164695.1 | NP_001158167.1 | 2 | 385 | |||
NM_001318495.1 | NP_001305424.1 | 2 | 385 | |||
NM_203395.2 | NP_981932.1 | 2 | 385 | |||
NR_134655.1 | 2 | 385 | ||||
XM_006715478.3 | XP_006715541.1 | 2 | 423 | |||
XM_006715479.3 | XP_006715542.1 | 2 | 284 | |||
GenBank mRNA | AK129950.1 | - | 2 | 215 | ||
AY259176.1 | - | 2 | 385 | |||
AY259177.1 | - | 2 | 385 | |||
AY424901.1 | - | 2 | 280 | |||
AY424902.2 | - | 2 | 280 | |||
AY957659.1 | - | 2 | 324 | |||
AY957660.1 | - | 2 | 324 | |||
AY957661.1 | - | 2 | 103 | |||
AY957662.1 | - | 2 | 122 | |||
BC056253.1 | - | 2 | 269 | 4612526 |