Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LAIR2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_002288.5 | NP_002279.2 | 5 | 633 | ||
NM_021270.4 | NP_067154.1 | 4 | 582 | |||
XM_011526961.2 | XP_011525263.1 | 4 | 584 | |||
GenBank mRNA | AA133246.1 | - | 5 | 631 | ||
AF013250.1 | - | 4 | 483 | |||
BX106365.1 | - | 5 | 633 |